MilliporeSigma
HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND I 2000
HPLC Analysis of Hydrobenzoin Enantiomers on Astec® CYCLOBOND I 2000 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Product No.
Description
Pricing
meso-Hydrobenzoin
99%
(S,S)-(−)-Hydrobenzoin
99%, optical purity ee: 99% (GLC)
(R,R)-(+)-Hydrobenzoin
99%, optical purity ee: 99% (HPLC)
Hydrobenzoin
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] 20 mM ammonium acetate, pH 4.1; [B] acetonitrile; (80:20, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null