MilliporeSigma
HPLC Analysis of Benzoin Enantiomers on Astec® (R,R) P-CAP-DP
HPLC Analysis of Benzoin Enantiomers on Astec® (R,R) P-CAP™-DP application for HPLC
Materials
Standard
Product No.
Description
Pricing
(S)-(+)-Benzoin
99%
Benzoin
98%
Benzoin
purified by sublimation, ≥99.5%
(R)-(−)-Benzoin
98%
CONDITIONS
column
Astec (R,R) P-CAP-DP, 25 cm x 4.6 mm I.D., 5 μm particles (35024AST)
mobile phase
[A] hexane; [B] ethanol; (80:20, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 230 nm
injection
2 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
P-CAP is a trademark of Sigma-Aldrich Co. LLC