MilliporeSigma
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection)
HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R (MS Detection) application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® R Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 2.1 mm
Application
Product No.
Description
Pricing
LC/MS Analysis of Cannabinoids on Ascentis® RP-Amide
application for HPLC
Standard
Product No.
Description
Pricing
Ketoprofen
meets USP testing specifications
Ketoprofen
≥98% (TLC)
(S)-(+)-Ketoprofen
99%
Ketoprofen
VETRANAL®, analytical standard
CONDITIONS
column
CHIROBIOTIC R, 15 cm x 2.1 mm, 5 μm particles (13019AST)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6 [B] methanol; (70:30, A:B)
flow rate
0.2 mL/min
column temp.
35 °C
detector
ESI(-)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC