MilliporeSigma
HPLC Analysis of Carotene Compounds on Ascentis® RP-Amide
HPLC Analysis of Carotene Compounds on Ascentis® RP-Amide application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® RP-Amide HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 μm particles (565324-U)
mobile phase
100% acetonitrile
flow rate
1.0 mL/min
column temp.
35 °C
detector
MS, ESI(+) in selective ion monitoring (SIR) mode
injection
5 μL
sample
10 μg/mL each in 100% acetonitrile
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany