HPLC Analysis of Carboxymethylated Apohemoglobin (Horse) Tryptic Digest on Discovery® BIO Wide Pore C18
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm × 2.1 mm I.D., 3 μm particles (568222-U)
mobile phase
[A] 0.1% TFA in water; [B] 0.1% TFA in water:0.1% TFA in acetonitrile, 50:50;
gradient
0-100% B in 50 min
flow rate
0.2 mL/min
column temp.
35°C
detector
UV, 220 nm
injection
5 μL, each at 0.8 mg/mL in 0.1% TFA
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany