MilliporeSigma
GC Analysis of Volatiles in White Wine on VOCOL® after SPME using a 75 μm Carboxen/PDMS Fiber
GC Analysis of Volatiles in White Wine on VOCOL® after SPME using a 75 μm Carboxen/PDMS Fiber suitable for GC, application for SPME
Materials
Analytical column
Product No.
Description
Pricing
VOCOL® Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 1.50 μm
Related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with manual sampling
SPME Fiber Holder
for use with CTC CombiPAL and Gerstel MPS2
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS)
df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Standard
Product No.
Description
Pricing
Hexyl acetate
analytical standard
Octanoic acid
analytical standard
Ethyl butyrate
analytical standard
Isoamyl alcohol
≥98%, FG
2-Methyl-1-butanol
≥99%
Sulfur dioxide
≥99.9%
Ethyl hexanoate
analytical standard
Ethyl octanoate
analytical standard
2,3-Butanediol
analytical standard, mixture of racemic and meso forms
mobile phase component
Product No.
Description
Pricing
Methyl formate
analytical standard
CONDITIONS
sample/matrix
white wine + 25% NaCl
SPME fiber
Carboxen/PDMS, 75 μm (57318)
extraction
10 min, 40 °C (headspace)
desorption process
3 min at 290 °C
column
VOCOL, 30 m × 0.25 mm I.D., 1.5 μm (24205-U)
oven
40 °C (2 min) to 220 °C at 10 °C/min
inj. temp.
290 °C
detector
GC-MS, quadrupole, m/z = 31-240
carrier gas
helium, 35 cm/sec
injection
splitless/split, closed 1 min
liner
0.75 mm liner
sample
white wine
Description
Analysis Note
In this application, SPME was used for the analysis of volatiles in white wine using a headspace method. The presence of aroma compounds in white wine is one of the most important attributes of its final quality. A Carboxen/PDMS fiber was inserted into the headspace of the vial for 10 minutes at 40 °C and analyzed by GC/MS. The thick film of the VOCOL GC column helps focus the volatile analytes.
Legal Information
VOCOL is a registered trademark of Merck KGaA, Darmstadt, Germany