MilliporeSigma
GC Analysis of α-Bromo-γ-Butyrolactone Enantiomers on Astec® CHIRALDEX G-DP
GC Analysis of α-Bromo-γ-Butyrolactone Enantiomers on Astec® CHIRALDEX™ G-DP suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-DP Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
standard
Product No.
Description
Pricing
α-Bromo-γ-butyrolactone
97%
CONDITIONS
column
Astec CHIRALDEX G-DP, 30 m x 0.25 mm I.D., 0.12 μm (78033AST)
oven
160 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
sample
peaks 1 & 2: α-bromo-γ-butyrolactone enantiomers
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC