Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

WH0000931M1

Sigma-Aldrich

Monoclonal Anti-MS4A1 antibody produced in mouse

clone 5C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-B1, Anti-Bp35, Anti-CD20, Anti-LEU16, Anti-MGC3969, Anti-MS4A2, Anti-S7, Anti-membrane-spanning 4-domains, subfamily A, member 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5C11, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MS4A1(931)

General description

This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This gene encodes a B-lymphocyte surface molecule which plays a role in the development and differentiation of B-cells into plasma cells. This family member is localized to 11q12, among a cluster of family members. Alternative splicing of this gene results in two transcript variants which encode the same protein. (provided by RefSeq)

Immunogen

MS4A1 (AAH02807, 210 a.a. ~ 297 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
GIVENEWKRTCSRPKSNIVLLSAEEKKEQTIEIKEEVVGLTETSSQPKNEEDIEIIPIQEEEEEETETNFPEPPQDQESSPIENDSSP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Ciccarelli et al.
The Lancet. Neurology, 13(8), 807-822 (2014-07-11)
The mechanisms underlying the pathogenesis of multiple sclerosis induce the changes that underpin relapse-associated and progressive disability. Disease mechanisms can be investigated in preclinical models and patients with multiple sclerosis by molecular and metabolic imaging techniques. Many insights have been
O Aubert et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(6), 1439-1445 (2014-05-09)
Anti-HLA donor-specific antibodies (DSAs) cause acute and chronic antibody-mediated rejection (AMR). However, the clinical relevance of anti-HLA-C antibodies remains unclear. We evaluated the clinical relevance of the presence of anti-HLA-C DSA at day 0 in renal transplant recipients. In this
Lisa G Rider et al.
Clinical and experimental rheumatology, 32(5), 689-696 (2014-07-30)
We aimed to assess changes in myositis core set measures and ancillary clinical and laboratory data from the National Institutes of Health's subset of patients enrolled in the Rituximab in Myositis trial. Eighteen patients (5 dermatomyositis, 8 polymyositis, 5 juvenile
Catalina Lee-Chang et al.
Blood, 124(9), 1450-1459 (2014-07-20)
Although the accumulation of highly-differentiated and granzyme B (GrB)-expressing CD8(+)CD28(-) T cells has been associated with aging, the mechanism for their enrichment and contribution to immune function remains poorly understood. Here we report a novel B-cell subset expressing 4-1BBL, which
Josef Singer et al.
Molecular cancer therapeutics, 13(7), 1777-1790 (2014-04-24)
Passive immunotherapy with monoclonal antibodies represents a cornerstone of human anticancer therapies, but has not been established in veterinary medicine yet. As the tumor-associated antigen EGFR (ErbB-1) is highly conserved between humans and dogs, and considering the effectiveness of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service