SAB5200656
Monoclonal Anti-Dicer - Atto 488 antibody produced in mouse
clone S167-7, purified immunoglobulin
Synonym(s):
Anti-Double-strand-specific ribonuclease, Anti-HERNA, Anti-KIAA0928
About This Item
biological source
mouse
Quality Level
conjugate
Atto 488 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S167-7, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 215 kDa
species reactivity
human, mouse, rat
concentration
1 mg/mL
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Dicer1(192119)
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
SKU | GTIN |
---|---|
SAB5200656-100UG | 4061832148564 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service