Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700873

Sigma-Aldrich

Monoclonal Anti-OPAL1 antibody produced in mouse

clone OPAL1-01

Synonym(s):

WBP1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
OPAL1-01, monoclonal
application:
FACS
species reactivity:
human
technique(s):
flow cytometry: 2 μg/mL
citations:

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

OPAL1-01, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: 2 μg/mL

accession no.

Q9NX94

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... WBP1L(54838)

General description

The mouse monoclonal antibody OPAL1-01 recognizes OPAL1 (outcome predictor in acute leukemia 1), a transmembrane adaptor protein expressed mainly by megacaryocytes and platelets.

Immunogen

Recombinant fragment of human OPAL1 (amino acids 152-342)

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service