Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EHU159581

Sigma-Aldrich

MISSION® esiRNA

targeting human WIPF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGATCCACATCTGCGAAACCCTTTTCACCCCCAAGTGGCCCAGGGAGGTTTCCTGTGCCTTCTCCAGGCCACAGAAGTGGTCCCCCAGAGCCTCAGAGGAACCGAATGCCGCCCCCAAGGCCCGACGTGGGCTCAAAGCCTGATAGCATTCCTCCTCCAGTACCTAGTACTCCAAGACCCATTCAATCAAGTCCGCACAAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Pan et al.
Journal of experimental & clinical cancer research : CR, 37(1), 167-167 (2018-07-26)
Aberrant expression of Wiskott-Aldrich syndrome protein interacting protein family member 1 (WIPF1) contributes to the invasion and metastasis of several malignancies. However, the role of WIPF1 in human pancreatic ductal adenocarcinoma (PDAC) remains poorly understood. Human pancreatic cancer samples from
Amrita Salvi et al.
Biochemical and biophysical research communications, 482(4), 1353-1359 (2016-12-13)
Cancer cell migration and invasion involves actin cytoskeleton reorganization, which is regulated by the WASP (Wiskott Aldrich Syndrome Protein) family of proteins such as WASP, N-WASP (Neural-WASP) and WASP interacting protein (WIP). In this study, we found that the expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service