Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

663468

Sigma-Aldrich

Rhodium on alumina

extent of labeling: 5 wt. % loading, Degussa type G214 RA/D

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Quality Level

form

powder

reaction suitability

core: rhodium
reagent type: catalyst

extent of labeling

5 wt. % loading

SMILES string

[Rh]

InChI

1S/Rh

InChI key

MHOVAHRLVXNVSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for hydrogenation of unsaturated compounds including aldehydes, ketones, andaromatic carbocyclic and heterocyclic structures; high stereospecificity and low hydrogenolytic activity".

Storage Class

13 - Non Combustible Solids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service