材料
black phenolic cap
特點
closure type black phenolic hole cap
包裝
pkg of 200 ea
接頭
thread for 20-400
正在寻找类似产品? 访问 产品对比指南
一般說明
It is autoclavable, easy sterilization, is used as a closure type for the containers used to contain substances in order to prevent contamination.
Global Trade Item Number
货号 | GTIN |
---|---|
Z127299-200EA | 4061826727423 |