MilliporeSigma
HPLC Analysis of Chloroquine Enantiomers on Astec® CHIROBIOTIC® V
HPLC Analysis of Chloroquine Enantiomers on Astec® CHIROBIOTIC® V application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Standard
Product No.
Description
Pricing
Chloroquine diphosphate salt
powder or crystals, 98.5-101.0% (EP)
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine; (98:1:1, A:B:C)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 220 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC