MilliporeSigma
HPLC Analysis of Pyroglutamic Acid Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of Pyroglutamic Acid Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] acetonitrile; [B] methanol; [C] acetic acid; [D] triethylamine; (54.5:45:0.3:0.2, A:B:C:D)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 220 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC