MilliporeSigma
HPLC Analysis of Proteins on Mono Q HR 5/5
HPLC Analysis of Proteins on Mono Q HR 5/5 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Mono Q® 5/50 GL
10 μm particle size, L × I.D. 5 cm × 5 mm
CONDITIONS
column
Mono Q HR 5/5, 10 μm particles (54807)
mobile phase
[A]: 20 mM bis-Tris, pH 6.0; [B]: A + 1M sodium chlorde
flow rate
1.0 mL/min
detector
UV, 280 nm
injection
2.5 mL
sample
human placental lactogen fraction from size exclusion medium