MilliporeSigma
HPLC Analysis of 4-Phenyl-2-oxazolidinone Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of 4-Phenyl-2-oxazolidinone Enantiomers on Astec® CHIROBIOTIC® T application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® T Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
(S)-(+)-4-Phenyl-2-oxazolidinone
98%
(R)-(−)-4-Phenyl-2-oxazolidinone
98%
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] 0.5% triethylamine acetate, pH 4.1; [B] methanol; (80:20, A:B)
flow rate
1 mL/min
column temp.
ambient
detector
UV, 254 nm
injection
5 μL
sample
5 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC