MilliporeSigma
Standard Methods 6040D: GC Analysis of Geosmin and 2-MIB on SLB®-5ms after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber
Standard Methods 6040D: GC Analysis of Geosmin and 2-MIB on SLB®-5ms after SPME using 50/30 μm DVB/Carboxen/PDMS Fiber suitable for GC, application for SPME
Materials
Analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS)
for use with autosampler, needle size 23 ga, metal alloy fiber, fiber L 2 cm
CONDITIONS
SPME fiber
2 cm Metal coated with 50/30 μm DVB/Carboxen/PDMS (57914-U)
extraction
headspace, 65 °C (30 min)
desorption process
260 °C for 3 min.
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
60 °C (2 min), 8 °C/min to 200 °C
MSD interface
300 °C
scan range
SIM, m/z = 137, 124, 95, 112
carrier gas
helium, 1 mL/min constant
liner
0.75 mm I.D., SPME
sample
20/10 ppt odor compounds in 25 mL water + 25% NaCl
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany