GC Analysis of Geranium Bourbon Essential Oil on SUPELCOWAX 10
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
50 °C (2 min), 2 °C/min to 230 °C (1 min)
detector
FID
carrier gas
helium, 20 cm/sec @ 155 °C
injection
0.2 μL, 100:1 split
sample
neat oil
Description
Legal Information
null