MilliporeSigma
  • The role of RIP3 mediated necroptosis in ouabain-induced spiral ganglion neurons injuries.
The role of RIP3 mediated necroptosis in ouabain-induced spiral ganglion neurons injuries.
Neuroscience letters (2014-07-06)
Xi Wang, Ye Wang, Zhong-jia Ding, Bo Yue, Peng-zhi Zhang, Xiao-dong Chen, Xin Chen, Jun Chen, Fu-quan Chen, Yang Chen, Ren-feng Wang, Wen-juan Mi, Ying Lin, Jie Wang, Jian-hua Qiu
ABSTRACT
Spiral ganglion neuron (SGN) injury is a generally accepted precursor of auditory neuropathy. Receptor-interacting protein 3 (RIP3) has been reported as an important necroptosis pathway mediator that can be blocked by necrostatin-1 (Nec-1). In our study, we sought to identify whether necroptosis participated in SGN injury. Ouabain was applied to establish an SGN injury model. We measured the auditory brain-stem response (ABR) threshold shift as an indicator of the auditory conditions. Positive β3-tubulin immunofluorescence staining indicated the surviving SGNs. RIP3 expression was evaluated using immunofluorescence, quantitative real-time polymerase chain reaction and western blot. SGN injury promoted an increase in RIP3 expression that could be suppressed by application of the necroptosis inhibitor Nec-1. A decreased ABR threshold shift and increased SGN density were observed when Nec-1 was administered with apoptosis inhibitor N-benzyloxycarbonyl-Val-Ala-Asp-fluoromethylketone (Z-VAD). These results demonstrated that necroptosis is an indispensable pathway separately from apoptosis leading to SGN death pathway, in which RIP3 plays an important role.
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)