Skip to Content
MilliporeSigma
N-Acetyl-L-cysteine BioXtra, ≥99% (TLC)

N-Acetyl-L-cysteine

N-Acetyl-L-cysteine BioXtra, ≥99% (TLC)
Synonym(s):
LNAC, NAC, N-Acetyl-L-cysteine
Linear Formula:
HSCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
163.19
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Optical Isomer
Amino Acid
Technique