About This Item
Kod UNSPSC:
41122801
NACRES:
NB.01
Polecane produkty
opakowanie
pkg of 1 ea
producent / nazwa handlowa
Finn 9420290
Finn
Opis ogólny
Rugged molded supports provide plenty of height for the longest Finnpipettes. Aluminum crossbar has curved upper edge to fit securely under the finger support of any Finnpipette single-channel or multi-channel pipette.
Informacje prawne
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)