Kluczowe dokumenty
Z329703
BRAND® wash bottle, wide-mouth, labelled, LDPE
capacity 250 mL, label, Ethanol, without venting valve
About This Item
Kod UNSPSC:
41103209
NACRES:
NB.17
Materiały
low-density polyethylene bottle
orange closure
round bottle
translucent bottle
wide-mouth bottle
Właściwości
label Ethanol
with cap
without venting valve
opakowanie
pack of 5 ea
producent / nazwa handlowa
BRAND 144082
pojemność
250 mL
Opis ogólny
Flexible, high-quality wash bottles with integral wash steam and sealing cone in the closure. Large opening for convenient filling. Label with designation of contents and general safety statements (with CAS no. and NFPA codes).
Bottles with venting valve (metal ball) are for distilled water and organic solvents only; to avoid dropping caused by overpressure.
Inne uwagi
BR144082 (Indie)
Informacje prawne
BRAND is a registered trademark of BRAND GMBH + CO KG
wyposażenie dodatkowe
Numer produktu
Opis
Cennik
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)