Kluczowe dokumenty
V0756
Sigma-Aldrich
Vortex adapter for microcentrifuge tubes
capacity, 20 tubes
About This Item
Kod UNSPSC:
41121702
eCl@ss:
32119210
NACRES:
NB.73
Właściwości
capacity 20 tubes
opakowanie
pack of 1 ea
producent / nazwa handlowa
Bel-Art H37080-0020
Opis ogólny
For use with any vortex mixer equipped with a rubber pad or test tube cup. The device holds 1.5 mL tubes in a disk mounted on a shaft. A sliding plate holds down the tube caps when in use. Provides rapid and complete mixing of many samples at once.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)