RA817-EP
Rotor for Eppendorf® 5702/5702R/5702RH Centrifuges
A-8-17, aerosol-tight, to hold, 8 x 15 mL conical tubes, without adapter, without rubber mats
About This Item
Polecane produkty
Właściwości
to hold 8 x 15 mL conical tubes
aerosol-tight
low-capacity swing bucket rotor
without adapter
without rubber mats
opakowanie
pack of 1 ea
producent / nazwa handlowa
Eppendorf® 022639501 (NA)
Eppendorf® 5702700002 (EU)
Parametry
2,800 × g max. RCF
metody
centrifugation: suitable
kompatybilność
for use with centrifuge models 5702/5702R/5702RH