About This Item
Kod UNSPSC:
41105101
eCl@ss:
32012303
NACRES:
NB.24
Polecane produkty
Warunki transportu
ambient
Opis ogólny
This item includes parts for catalog numbers WP6111560, WP6122050, WP6110060
This item is a kit composed of the following items:
(1) Diaphragm hold-down plate assembly, (1) Valve plate assembly, (1) Pump head, (1) Vacuum gauge, (1) Pressure gauge, (4) Head screws, (2) Handle screws
This item is a kit composed of the following items:
(1) Diaphragm hold-down plate assembly, (1) Valve plate assembly, (1) Pump head, (1) Vacuum gauge, (1) Pressure gauge, (4) Head screws, (2) Handle screws