AIRGAPXL2
Air Gap Device
For use with Milli-Q® CLX, HX, and HR 7000 series water purification systems.
About This Item
Kod UNSPSC:
41104200
NACRES:
JA.13
Polecane produkty
opakowanie
pkg of 1 unit
kompatybilność
for use with Milli-Q® CLX
for use with Milli-Q® HR
for use with Milli-Q® HX
Opis ogólny
This accessory can be connected directly to a 10 mm OD tubing to provide protection from bacterial contamination on the reject tubing.
Zastosowanie
For the bacteriological protection of reject lines.
Informacje prawne
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany