Z764590
Stopcock Adapter
2 mm PTFE stopcock, bent, joint: ST/NS 29/32
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Polecane produkty
Połączenie
joint: ST/NS29/32
Opis ogólny
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)