Z562068
Platinum crucible lid
fits 15 mL crucible
Synonim(y):
crucibles
About This Item
Kod UNSPSC:
41122200
NACRES:
NB.43
Polecane produkty
producent / nazwa handlowa
Birmingham metals
masa
3.6 g
Opis ogólny
Platinum labware has excellent resistance to acids such as acetic, hydrochloric, hydrofluoric, hydroiodic, nitric, selenic, and sulfuric, and to oxidation at high temperatures. Crucible lids of platinum can be cleaned by boiling in a suitable solvent.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
For the removal of substances which have alloyed with the surface, scour with alumina powder.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z562068-1EA | 4061833310694 |