Z407186
Aldrich® Sure/Seal™ septum-inlet transfer adapter
polypropylene, pkg of 1 ea
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.43
Polecane produkty
Materiały
polypropylene
opakowanie
pkg of 1 ea
Powiązane kategorie
Opis ogólny
This inexpensive septum-inlet adapter screws over the Sure/Seal crown to permit repeated dispensing of product via syringe while protecting the contents of the bottle from air and moisture. The adapter allows either an 8 mm septum cap or standard rubber septa to be used.
The 8 mm septum cap (included) with flat PTFE-faced septum screws onto the side-arm for dispensing or short term storage. Standard rubber septa (Z565695 or Z565679) fit on side-arms for dispensing. For use with Aldrich products packed in Sure/Seal bottles.
The 8 mm septum cap (included) with flat PTFE-faced septum screws onto the side-arm for dispensing or short term storage. Standard rubber septa (Z565695 or Z565679) fit on side-arms for dispensing. For use with Aldrich products packed in Sure/Seal bottles.
Powiązanie
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z421537-1PAK | 4061832966915 |
Z407186-1EA | 4061837619687 |
Z407186-5EA |