Z286079
Ace vacuum manifold system
#15 Ace-Threds
About This Item
Kod UNSPSC:
40161501
NACRES:
NB.79
Polecane produkty
producent / nazwa handlowa
Ace Glass
Powiązane kategorie
Opis ogólny
Set includes size-matched components with #15 Ace-Threds for easy assembly of vacuum system. Order 1/2 in. I.D. vacuum tubing separately.
- 1 Vacuum manifold, dual bank, 3-ports with 5-ring medium hose barb connections, and greaseless, 0 to 4 mm, double action, high-vacuum PTFE stopcocks
- 1 Mineral oil bubbler, adjustable
- 1 Vacuum trap, adjustable
- 4 UHDPE tubing connectors for ½ in. I.D. vacuum tubing
- 2 Nylon plugs
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Klienci oglądali również te produkty
Global Trade Item Number
SKU | GTIN |
---|---|
Z286079-1EA | 4061832363158 |