Z278785
Bytac® surface protection laminate
size × thickness 12 1/2 in. × 15 ft × 0.003 mm
Synonim(y):
Laminat stołowy, ochrona powierzchni roboczej, ochrona ławki
About This Item
Kod UNSPSC:
41122300
NACRES:
NB.35
Polecane produkty
Materiały
aluminum backing
opakowanie
pack of 1 ea
rozmiar × grubość
12 1/2 in. × 15 ft × 0.003 mm
Powiązane kategorie
Opis ogólny
Add the benefits of FEP to any flat surface. Bytac protects surfaces from absorption of toxic and radioactive materials. The laminate consists of FEP resin supported on a backing of vinyl or aluminum. The backing has a pressure sensitive acrylic adhesive that adheres to most surfaces. Vinyl backing is recommended for applications up to 200 °F (93 °C). Aluminum backing is for high-temperature applications and can be used up to 360 °F (182 °C).
- non-stick; easy to clean
- chemically inert
Informacje prawne
Bytac is a registered trademark of Saint-Gobain Performance Plastics Corp.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z278785-1EA | 4061833569030 |