Z266213
Polymer-body combination pH electrodes
BNC fitting
About This Item
Kod UNSPSC:
41115603
NACRES:
NB.79
Polecane produkty
trzon dł. × śr.
110 mm × 12 mm
Opis ogólny
General-purpose, double junction
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Hints on Electrode Maintenance
The combination of a pH indicating and reference half-cell in an impact-resistant epoxy body. Designed for applications where breakage is a problem.
- Environmental - field sampling
- Industrial - process control
- Educational - core courses
produkt powiązany
Numer produktu
Opis
Cennik
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z266213-1EA | 4061837617973 |