Key Documents
CLS53202L
Pyrex® heavy-wall graduated filtering flask
capacity 2,000 mL
Synonim(y):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
About This Item
Kod UNSPSC:
41121804
NACRES:
NB.43
Materiały
borosilicate glass
flat bottom flask
narrow-mouth
Połączenie
joint stopper9
opis
without hose connection
sterylność
non-sterile
Właściwości
closure type Rubber stopper
opakowanie
pack of 1
case of 12
producent / nazwa handlowa
Corning 5320-2L
zakres pomiaru objętości
600-2,000 mL
pojemność
2,000 mL
Opis ogólny
Blown in special molds to give maximum strength. Graduated to show approximate capacity.
Informacje prawne
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.