Key Documents
MIX0500L100-M
Millipore
Mobius® MIX500, 208 V, 50/60 Hz
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
41103801
eCl@ss:
32010600
AC/DC 입력
208 V, 50 - 60 Hz
Quality Level
성분
Includes carrier, control box unit, motor, hand-held control and cable
법적 정보
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany