Merck
Z740422
Tamper proof specimen containers
size 1000 mL
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121800
NACRES:
NB.22
物質
polypropylene cap
polypropylene container
包装
pack of 100 ea
メーカー/製品名
Simport C581-1L
サイズ
1000 mL
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
These disposable containers are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples. Tight-fitting lids prevent leaks and odors. They incorporate a tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover. They resist temperatures up to 80 °C. The two larger sizes are supplied with a plastic handle for easy carrying.
最新バージョンのいずれかを選択してください:
試験成績書(COA)