Merck
Z332445
BRAND® discharge tube for Dispensette®
with integrated valve, for 5 & 10 mL Dispensette III
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41121500
NACRES:
NB.01
メーカー/製品名
BRAND
L
90 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
関連するカテゴリー
詳細
The BRAND® Discharge Tube is designed for use with both the Dispensette® and Seripettor® systems, providing a reliable solution for precise liquid dispensing. This versatile discharge tube includes essential components such as a closure cap, EPDM discharge valve, and sealing ring, ensuring optimal performance and convenience.
その他情報
BR707916 (India)
法的情報
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
最新バージョンのいずれかを選択してください:
試験成績書(COA)