Merck
TRT51134
Extra Wide Compact Locking 4 Drawer Cart
purple (drawers)
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41123300
NACRES:
JA.01
物質
polyethylene
purple (drawers)
メーカー/製品名
TrippNT 51134
W × H × D
24 1/2 × 3 3/8 × 15 1/2 in., drawers
重量
67 lb
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
An extra-wide compact cart with 4 drawers; fits under most counters, tables, desks and work benches.
  • Massive under counter storage
  • One piece construction
  • Non metal rust proof polymer construction
  • Decontaminates with bleach or aqueous solutions
  • Built-in tray top and handle for push pull control
  • White polyethylene body, ABS drawers
  • 13 colors
  • 4 full-extension drawers
  • Drawer headspace: 4 1/2 inches
  • 3 inch polypropylene swivel casters, 2 locking
  • American Made
最新バージョンのいずれかを選択してください:
試験成績書(COA)