Merck
G6778
Sigma-Aldrich
Gaskets for vertical standard gels, 16.5 cm wide
size 1.0 mm × 19 cm, color-coded red
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41105305
サイズ
1.0 mm × 19 cm
品質水準
color-coded red
類似した製品をお探しですか? 訪問 製品比較ガイド
特徴および利点
Unique silicone rubber gasket provides continuous seal around sides and bottom of glass plates, eliminating the need for a bottom spacer. Order gasket thickness to match comb and spacers. Gaskets are color-coded by thickness for easy identification. Length given is height of assembly; actual gasket length is 2 × h + w.