Merck
EP5895135005
Rotor for Eppendorf® 5920R Centrifuges
FA-48x2, aerosol tight, to hold, 48 x 1.5/2.0 mL tubes, with Eppendorf QuickLock rotor lid
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41103909
NACRES:
NB.82
特徴
to hold 48 x 1.5/2.0 mL tubes
aerosol tight
fixed angle rotor
with Eppendorf QuickLock rotor lid
包装
pack of 1 ea
メーカー/製品名
Eppendorf® 5895135005 (Global)
パラメーター
20,913 × g max. RCF
テクニック
centrifugation: suitable
適合性
for use with centrifuge models 5920R
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
法的情報
Eppendorf is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)