Merck
EP5391000036
Eppendorf(R) HeatSealer
S100
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41102400
NACRES:
NC.21
AC/DC入力
230 V, 50/60 Hz, UK plug
特徴
CE compliant
CSA approved
fixed Sealing, temperature
seals 96 and 384 well plates
メーカー/製品名
Eppendorf® 5391000036 (GB)
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
The Eppendorf HeatSealer Family offers you reliable and effective Sealing, of a variety of plates. You can choose from a more compact (S100) and a more flexible (S200) instrument according to your Sealing, needs.
特徴および利点
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
法的情報
Eppendorf is a registered trademark of Eppendorf AG
最新バージョンのいずれかを選択してください:
試験成績書(COA)