おすすめの製品
フォーム
powder (dehydrated (DCM))
品質水準
メーカー/製品名
GranuCult®
テクニック
microbiological culture: suitable
アプリケーション
food and beverages
microbiology
保管温度
15-25°C
適合性
Bacillus cereus
関連するカテゴリー
詳細
for microbiology
関連事項
Replaces: 1.01868.0107
アナリシスノート
Appearance (clearness): turbid with sediment
Appearance (colour): brownish
pH-value (25 °C): 6.6 - 7.0
Duopath Cereus NHE (Bacillus cereus MHI 26): +
Duopath Cereus NHE (Bacillus cereus ATCC 11778 (WDCM 00001)): +
Duopath Cereus NHE (Bacillus subtilis ATCC 6633 (WDCM 00003)): -
Duopath Cereus HBL (Bacillus cereus MHI 26): +
Duopath Cereus HBL (Bacillus cereus ATCC 11778 (WDCM 00001)): -
Duopath Cereus HBL (Bacillus subtilis ATCC 6633 (WDCM 00003)): -
Appearance (colour): brownish
pH-value (25 °C): 6.6 - 7.0
Duopath Cereus NHE (Bacillus cereus MHI 26): +
Duopath Cereus NHE (Bacillus cereus ATCC 11778 (WDCM 00001)): +
Duopath Cereus NHE (Bacillus subtilis ATCC 6633 (WDCM 00003)): -
Duopath Cereus HBL (Bacillus cereus MHI 26): +
Duopath Cereus HBL (Bacillus cereus ATCC 11778 (WDCM 00001)): -
Duopath Cereus HBL (Bacillus subtilis ATCC 6633 (WDCM 00003)): -
脚注
We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
法的情報
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
保管分類コード
11 - Combustible Solids
WGK
WGK 3
試験成績書(COA)
製品のロット番号・バッチ番号を入力して、試験成績書(COA) を検索できます。ロット番号・バッチ番号は、製品ラベルに「Lot」または「Batch」に続いて記載されています。
ライフサイエンス、有機合成、材料科学、クロマトグラフィー、分析など、あらゆる分野の研究に経験のあるメンバーがおります。.
製品に関するお問い合わせはこちら(テクニカルサービス)