Merck
BAH186100440
Stak-A-Tray system rack
for petri dishes and small culture flasks
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41122800
NACRES:
NB.35
物質
stainless steel
特徴
autoclavable
Petri dishes and small culture flasks
包装
pack of 1 ea
メーカー/製品名
Bel-Art H18610-0440
W × D × H
35.8 cm × 37.5 cm × 10.8 cm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Choose from two rack models; one stores two levels of trays to support taller vessels like tissue culture flasks, the other stores four levels of trays for shorter containers like dishes and plates. All racks interlock upon stacking for a small, space-saving footprint that holds up to 20 levels of trays. Every level supports one large or two small interchangeable trays, that are purchased separately. Air flows through perforations in the trays and every part is fabricated of stainless steel and is steam sterilizable at 121 °C (250 °F). Racks and trays sold separately.
  • Versatile configurations to accommodate many types of media plates and flasks
  • Autoclavable
法的情報
Stak-A-Tray is a trademark of SP Scienceware, a division of SP
最新バージョンのいずれかを選択してください:
試験成績書(COA)