Merck
BAF132040000
Cover for bio-hazard incinerator carton
autoclavable
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41100000
NACRES:
NB.35
特徴
autoclavable
包装
pack of 1 ea
メーカー/製品名
Bel-Art F13204-0000
詳細
Dishwasher-safe and autoclave safe, the reusable Disposal Carton Cover opens quickly and easily by pressing the built-in release tab – no batteries required! Fits any disposal carton that has a 30.5 x 30.5cm (12 x 12") opening.
  • Opens with a single touch
  • Sanitary and reusable
  • Provides a wide opening 20.3 x 24.1 cm (8 x 9 1/2 in.) permitting safer and more sanitary deposit of wastes into the container
  • Red center panel matches printing on Biohazard disposal cartons providing fast visual recognition of the waste stream
  • Features a smooth 10.1 x 10.1 cm (4 x 4 in.) area on top for applying labels
最新バージョンのいずれかを選択してください:
試験成績書(COA)