Documenti fondamentali
41910-U
Supelco
Model 226 Pressure Regulator
with gauge,
single-stage
About This Item
Codice UNSPSC:
41112500
NACRES:
SB.54
Materiali
aluminum body
Raccordi
CGA 180
applicazioni
environmental
Descrizione generale
  • Compact size and light weight design for easy transportability.
  • Elgiloy® diaphragm minimizes diffusion resistance and provides accurate and stable pressure control.
  • Sintered filter protects internal components and extends regulator service life.
Aluminum body with stainless steel diaphragm
• Inlet pressure rating 0-2000psig
• Delivery pressure 1-25psig
• CGA 180 / 1/8-inch NPT (F)
• Supplied with 1/8-inch Swagelok NPT to 1/8-inch tube
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)