Documenti fondamentali
20436
Supelco
Plastic Column Cap
for use with 1/16 in. O.D. column
About This Item
Codice UNSPSC:
41121800
Confezionamento
pkg of 100 ea
tecniche
gas chromatography (GC): suitable
Raccordi
female cap for 1/16 in. O.D. column (slip-on)
Compatibilità
for use with 1/16 in. O.D. column
Descrizione generale
These plastic caps will easily slip over the ends of packed GC columns to protect them during storage.
Applicazioni
Plastic column cup maybe used to close the immunoaffinity chromatography column used for separating cross-linked protein complexes adhering to P1 protein of Mycoplasma pneumonia.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
G Layh-Schmitt et al.
Microbiology (Reading, England), 146 ( Pt 3), 741-747 (2000-04-04)
Adherence of Mycoplasma pneumoniae to host cells requires several mycoplasmal membrane proteins and cytoskeleton-like proteins in addition to the adhesin P1, a transmembrane protein of 170 kDa. To analyse interactions of the P1 adhesin with other membrane proteins or with