Documenti fondamentali
EP0030108426
Eppendorf® DNA LoBind Tubes
capacity 2.0 mL, PCR clean, withstand, 30,000 x g, pack of 500 ea
Sinonimo/i:
Eppendorf® DNA LoBind tubes
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.15
Materiali
colorless
conical bottom
Descrizione
Low retention
Sterilità
non-sterile
Caratteristiche
DNase free
PCR clean
Confezionamento
pack of 500 ea
Capacità
2.0 mL
Compatibilità
suitable for PCR
Descrizione generale
Maximize nucleic acid recovery. Eppendorf LoBind Tubes signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules. The ideal solution for sample preparation and long-term storage of your precious nucleic acids
Caratteristiche e vantaggi
Signifcantly reduce sample-to-surface binding to ensure maximal recovery of DNA and RNA molecules
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)