Documenti fondamentali
EP0030108442
Eppendorf® Protein LoBind Tubes
PCR clean, withstand 18,000 x g, 1.5 mL, pack of 500 ea
Sinonimo/i:
Eppendorf® Protein LoBind tubes
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.15
Materiali
cap (push fit)
colorless
conical bottom
Descrizione
Low retention
Sterilità
non-sterile
Caratteristiche
DNase free
PCR clean
Confezionamento
pack of 500 ea
Produttore/marchio commerciale
Eppendorf® 0030108442
Capacità
1.5 mL
Compatibilità
suitable for PCR
Descrizione generale
Avoid losing valuable samples during centrifugation, incubation and storage! Rely on the excellent protection offered by the hinged Safe-Lock lid. It contains all of our experience from 50 years of continuous optimization and development. Trust the original Eppendorf Safe-Lock Tubes, because your samples deserve only the best.
Caratteristiche e vantaggi
Eppendorf Protein LoBind Tubes are specifically designed to ensure you get maximal protein recovery.
Note legali
Eppendorf is a registered trademark of Eppendorf AG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)