Documenti fondamentali
Z258717
Sigma-Aldrich
Ace vacuum-sublimation apparatus
Complete apparatus, H × I.D. 250 mm × 152 mm
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.76
Produttore/marchio commerciale
Ace Glass 802355
Alt. × diam. int.
250 mm × 152 mm
Categorie correlate
Descrizione generale
The Ace vacuum-sublimation apparatus features a greaseless O-ring sealed flange with a quick-release clamp. The top flange is ground flat, bottom mating flange has a groove for a CAPFE (fluororesin encapsulated silicone) O-ring. The complete apparatus includes an outer body, inner top, CAPFE O-ring, and quick-release clamp.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Global Trade Item Number
SKUGTIN
Z258717-1EA4061826659892
Z258733-1EA4061832349077