Z230855
Aldrich® Schlenk-type male joint cap
male joint: ST/NS 14/20
Sinonimo/i:
Aldrich® Schlenk-type male joint cap
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.43
Prodotti consigliati
Etichetta
L
Giunto
male joint: ST/NS14/20
Categorie correlate
Descrizione generale
Thermoplastic screw caps with Viton O-rings usable to 140°C. This stopper has a rounded top and fits where clearances are tight.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
I clienti hanno visto anche
Articoli
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.